missing translation for 'onlineSavingsMsg'
Learn More

Celgosivir hydrochloride, MedChemExpress

Product Code. 16496988
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
16496988 10 mg -
16473516 25 mg -
16463047 50 mg 50mg
16449537 5 mg -
16431599 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496988 Supplier Medchem Express Supplier No. HY16134A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Celgosivir hydrochloride (MBI 3253 hydrochloride; MDL 28574 hydrochloride; MX3253 hydrochloride) is an α-glucosidase I inhibitor; inhibits bovine viral diarrhoea virus (BVDV) with an IC50 of 1.27 μM in in vitro assay.

Celgosivir hydrochloride (MBI 3253 hydrochloride

Specifications

Kemisk navn eller materiale Celgosivir hydrochloride
Fysisk form Solid
CAS 141117-12-6
Farve Gray
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C12H22ClNO5
Til brug med (applikation) COVID-19-anti-virus
Synonym MBI 3253 hydrochlorideMDL 28574 hydrochlorideMX3253 hydrochloride
Opløselighedsinformation H2O : ≥ 100 mg/mL (338.11 mM) ∣DMSO : 100 mg/mL (338.11 mM; Need ultrasonic)
SMIL O[C@@H]1[C@]2([H])[C@@H](O)[C@H](O)[C@@H](OC(CCC)=O)CN2CC1.Cl[H]
Molekylvægt (g/mol) 295.76
Formel vægt 295.76
Procent renhed 95.0%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.