Få mere at vide
Cefuroxime Sodium Salt, TRC
Artikelnummer. 30138168
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138168 5 g
17982635 25 g
17973875 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138168 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2480505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56238-63-2
Molekylær formel C16 H15 N4 O8 S . Na
Molekylvægt (g/mol) 446.37
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(aminocarbonyl)oxy]methyl]-7-[[(2Z)-2-furanyl(methoxyimino)acetyl]amino]-8-oxo-, monosodium salt, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(aminocarbonyl)oxy]methyl]-7-[[2-furanyl(methoxyimino)acetyl]amino]-8-oxo-, monosodium salt, [6R-[6α,7β(Z)]]-, 640/359, Anaptivan, Biociclin, Biofurex, Bioxima, Cefamar, Cefoprim, Cefumax, Cefurex, Cefurin, Cefuroxime sodium, Cefuroxime sodium salt, Curocef, Curoxim, Duxima, Gibicef, Ipacef, Kefurox, Kesint, Lampsporin, Medoxim, Novocef, Sodium cefuroxime, Spectrazol, Spectrazole, Ultroxim, Zinacef
IUPAC navn sodium;(6R,7R)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMIL [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])COC(=O)N)\c3occc3
Tekniske data
Kemisk navn eller materiale Cefuroxime Sodium
Mængde 5 g
InChI formel InChI=1S/C16H16N4O8S.Na/c1-26-19-9(8-3-2-4-27-8)12(21)18-10-13(22)20-11(15(23)24)7(5-28-16(17)25)6-29-14(10)20;/h2-4,10,14H,5-6H2,1H3,(H2,17,25)(H,18,21)(H,23,24);/q;+1/p-1/b19-9-;/t10-,14-;/m1./s1
Formel vægt 446.0508