Få mere at vide
Cefuroxime sodium, MedChemExpress
Artikelnummer. 16412329
Encompass_Preferred
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
16412329 1 g
1 options
Artikelnummer. 16412329 Leverandør Medchem Express Leverandørnr. HYB1256
MedChemExpress Cefuroxime sodium is an orally active second-generation cephalosporin antibiotic with increased stability to β-lactamase. Cefuroxime sodium has a broad spectrum activity against Gram-positive and Gram-negative bacteria.
Cefuroxime sodium is an orally active second-generation cephalosporin antibiotic with increased stability to β-lactamase. Cefuroxime sodium has a broad spectrum activity against Gram-positive and Gram-negative bacteria.
Tekniske data
Kemisk navn eller materiale Cefuroxime sodium
Fysisk form Solid
CAS 56238-63-2
Farve White
Mængde 1 g
Renhedsgrad noter Research
Molekylær formel C16H15N4NaO8S
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : ≥ 100 mg/mL (224.03 mM) ∣H2O : 50 mg/mL (112.01 mM; Need ultrasonic)
SMIL O=C(C(N12)=C(COC(N)=O)CS[C@]2([H])[C@H](NC(/C(C3=CC=CO3)=N\OC)=O)C1=O)O[Na]
Molekylvægt (g/mol) 446.37
Formel vægt 446.37
Procent renhed 99.08%