Få mere at vide
Cefuroxime Sodium, LoGiCal
Artikelnummer. 30457862
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30457862 10 mg
1 options
Artikelnummer. 30457862 Leverandør LoGiCal Leverandørnr. LGCFOR1341.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cefuroxime Sodium
CAS 56238-63-2
Analyt- eller komponentnavne Cefuroxime Sodium
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C16 H15 N4 O8 S . Na
Certificeringer/compliance ISO 9001
Synonym Cefuroxime Axetil Imp. D (EP), Cefuroxime sodium, Sodium (6R,7R)-3-[(carbamoyloxy)methyl]-7-[[(Z)-(furan-2-yl)(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, 750/2, 640/3, 640/68, 640/1, 640/2, GSK682709A, SD, SC, SB, 750/359, SA, Cefuroxime Sodium (640/359), GSK682708A
InChI formel InChI=1S/C16H16N4O8S.Na/c1-26-19-9(8-3-2-4-27-8)12(21)18-10-13(22)20-11(15(23)24)7(5-28-16(17)25)6-29-14(10)20;/h2-4,10,14H,5-6H2,1H3,(H2,17,25)(H,18,21)(H,23,24);/q;+1/p-1/b19-9-;/t10-,14-;/m1./s1
Løsningstype Neat
SMIL [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])COC(=O)N)\c3occc3
IUPAC navn sodium;(6R,7R)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Industry Type Clinical Testing
Molekylvægt (g/mol) 446.37
Mængde 10 mg
Formel vægt 446.0508 g/mol