Få mere at vide
Cefuracetime, TRC
Artikelnummer. 30363012
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363012 1 g
30350945 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363012 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2480401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 39685-31-9
Molekylær formel C17 H17 N3 O8 S
Molekylvægt (g/mol) 423.4
Synonym Cefuroxime Sodium Imp. B (EP), (6R,7R)-7-[[(Z)-(Furan-2-yl)(methoxyimino)acetyl]amino]-3-(hydroxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[[(2Z)-2-furanyl(methoxyimino)acetyl]amino]-8-oxo-, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[[2-furanyl(methoxyimino)acetyl]amino]-8-oxo-, [6R-[6α,7β(Z)]]-, (6R,7R)-3-[(Acetyloxy)methyl]-7-[[(2Z)-2-furanyl(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, Cefuracetime, SKF 81367
IUPAC navn (6R,7R)-3-(acetyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMIL CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)COC(=O)C)\c3occc3
Tekniske data
Kemisk navn eller materiale Cefuracetime
Mængde 1 g
InChI formel InChI=1S/C17H17N3O8S/c1-8(21)28-6-9-7-29-16-12(15(23)20(16)13(9)17(24)25)18-14(22)11(19-26-2)10-4-3-5-27-10/h3-5,12,16H,6-7H2,1-2H3,(H,18,22)(H,24,25)/b19-11-/t12-,16-/m1/s1
Formel vægt 423.0736