missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceftriaxone-d3 Disodium Salt, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Ceftriaxone O-methyl-d3 |
| CAS | 1132650-38-4 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C18 2H3 H15 N8 O7 S3 |
| Mængde | 10 mg |
| Synonym | 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxy-d3-imino)acetyl]amino]-8-oxo-3-[[(1,2,5,6-tetrahydro-2-methyl-5,6-dioxo-1,2,4-triazin-3-yl)thio]methyl]-, (6R,7R)-, (6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-(methoxy-d3-imino)acetyl]amino]-8-oxo-3-[[(1,2,5,6-tetrahydro-2-methyl-5,6-dioxo-1,2,4-triazin-3-yl)thio]methyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, Ceftriaxone-d3, Ceftriaxone-D3 (O-methyl-D3) |
| InChI formel | InChI=1S/C18H18N8O7S3/c1-25-18(22-12(28)13(29)23-25)36-4-6-3-34-15-9(14(30)26(15)10(6)16(31)32)21-11(27)8(24-33-2)7-5-35-17(19)20-7/h5,9,15H,3-4H2,1-2H3,(H2,19,20)(H,21,27)(H,23,29)(H,31,32)/b24-8-/t9-,15-/m1/s1/i2D3 |
| SMIL | [2H]C([2H])([2H])O\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CSC3=NC(=O)C(=NN3C)O)\c4csc(N)n4 |
| IUPAC navn | (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(trideuteriomethoxyimino)acetyl]amino]-3-[(6-hydroxy-2-methyl-5-oxo-1,2,4-triazin-3-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
| Molekylvægt (g/mol) | 557.6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?