missing translation for 'onlineSavingsMsg'
Learn More
Ceftizoxime, MedChemExpress Product Code.: 16498786

Ceftizoxime, MedChemExpress

Product Code. p-200098239
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16498786

Brand: Medchem Express HYB1596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ceftizoxime is a bacterial inhibitor which acts by interfering with bacterial cell wall synthesis and inhibiting cross-linking of the peptidoglycan.

Ceftizoxime is a bacterial inhibitor which acts by interfering with bacterial cell wall synthesis and inhibiting cross-linking of the peptidoglycan.

Specifications

Chemical Name or Material Ceftizoxime
CAS 68401-81-0
Color White
Purity Grade Notes Research
Molecular Formula C13H13N5O5S2
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (65.21 mM; Need ultrasonic)
SMILES O=C(C(N12)=CCS[C@]2([H])[C@H](NC(/C(C3=CSC(N)=N3)=N\OC)=O)C1=O)O
Molecular Weight (g/mol) 383.4
Formula Weight 383.4
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Ceftizoxime, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.