Få mere at vide
Ceftiofur Thiolactone, TRC
Artikelnummer. 30367952
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367952 2.5 mg
30361262 25 mg
30362615 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367952 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2447302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120882-23-7
Molekylær formel C14 H13 N5 O4 S3
Molekylvægt (g/mol) 411.479
IUPAC navn (2Z)-2-(2-amino-1,3-thiazol-4-yl)-N-[(4R,5R)-3,11-dioxo-6,10-dithia-2-azatricyclo[6.3.0.02,5]undec-1(8)-en-4-yl]-2-methoxyiminoacetamide
SMIL CO\N=C(/C(=O)N[C@H]1[C@H]2SCC3=C(N2C1=O)C(=O)SC3)\c4csc(N)n4
Tekniske data
Kemisk navn eller materiale Ceftiofur Thiolactone
Mængde 2.5 mg
InChI formel InChI=1S/C14H13N5O4S3/c1-23-18-7(6-4-26-14(15)16-6)10(20)17-8-11(21)19-9-5(2-24-12(8)19)3-25-13(9)22/h4,8,12H,2-3H2,1H3,(H2,15,16)(H,17,20)/b18-7-/t8-,12-/m1/s1
Formel vægt 411.013