Få mere at vide
Cefpodoxime Proxetil Isopropylcarbamate, TRC
Artikelnummer. 30364626
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364626 10 mg
30357049 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30364626 Leverandør Toronto Research Chemicals Leverandørnr. TRCC24387510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1192365-87-9
Molekylær formel C25H33N5O11S2
Molekylvægt (g/mol) 643.69
Synonym (6R,7R)-7-[[(2Z)-2-(Methoxyimino)-2-[2-[[(1-methylethoxy)carbonyl]amino]-4-thiazolyl]acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid 1-[[(1-Methylethoxy)carbonyl]oxy]ethyl Ester
IUPAC navn 1-propan-2-yloxycarbonyloxyethyl (6R,7R)-7-[[(2E)-2-methoxyimino-2-[2-(propan-2-yloxycarbonylamino)-1,3-thiazol-4-yl]acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMIL COCC1=C(N2[C@H](SC1)[C@H](NC(=O)\C(=N/OC)\c3csc(NC(=O)OC(C)C)n3)C2=O)C(=O)OC(C)OC(=O)OC(C)C
Tekniske data
Kemisk navn eller materiale Cefpodoxime Proxetil Isopropylcarbamate
Mængde 10 mg
InChI formel InChI=1S/C25H33N5O11S2/c1-11(2)38-24(34)28-23-26-15(10-43-23)16(29-37-7)19(31)27-17-20(32)30-18(14(8-36-6)9-42-21(17)30)22(33)40-13(5)41-25(35)39-12(3)4/h10-13,17,21H,8-9H2,1-7H3,(H,27,31)(H,26,28,34)/b29-16+/t13?,17-,21-/m1/s1
Formel vægt 643.1618