Få mere at vide
Thermo Scientific™ Cefoxitin, 98%
Brand: Thermo Scientific™ J66891.03
| Mængde | 1g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsCefoxitin is a semisynthetic, broad-spectrum second-generation cephalosporin with antibacterial activity. The activity of cefoxitin results in the weakening of the bacterial cell wall and causes cell lysis. Cefoxitin acts by interfe
Tekniske data
| Cefoxitin | |
| White | |
| >110°C (230°F) | |
| MFCD00072014 | |
| Predicted solubility in water is less than 0.2mg/mL | |
| COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)O)NC(=O)CC3=CC=CS3 | |
| 427.446 | |
| CHEBI:209807 | |
| 98% |
| 35607-66-0 | |
| 1g | |
| C16H17N3O7S2 | |
| 843°C | |
| WZOZEZRFJCJXNZ-ZBFHGGJFSA-N | |
| (6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
| 441199 | |
| 427.45 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.