Få mere at vide
Cefotiam Hexetil Hydrochloride (mixture of Diastereomers), TRC
Artikelnummer. 30364269
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364269 100 mg
30353086 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30364269 Leverandør Toronto Research Chemicals Leverandørnr. TRCC243000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cefotiam Hexetil hydrochloride (mixture of diastereomers)
CAS 95789-30-3
Molekylær formel C27 H37 N9 O7 S3 . 2 Cl H
Mængde 100 mg
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[2-(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, hydrochloride (1:2), (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, dihydrochloride, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, dihydrochloride, [6R-(6α,7β)]-, Cefotiam hexetil hydrochloride, Pansporin T, SCE 2174
InChI formel InChI=1S/C27H37N9O7S3.2ClH/c1-15(42-27(40)43-18-7-5-4-6-8-18)41-24(39)21-16(13-46-26-31-32-33-35(26)10-9-34(2)3)12-44-23-20(22(38)36(21)23)30-19(37)11-17-14-45-25(28)29-17;;/h14-15,18,20,23H,4-13H2,1-3H3,(H2,28,29)(H,30,37);2*1H/t15?,20-,23-;;/m1../s1
SMIL Cl.Cl.CC(OC(=O)OC1CCCCC1)OC(=O)C2=C(CSc3nnnn3CCN(C)C)CS[C@@H]4[C@H](NC(=O)Cc5csc(N)n5)C(=O)N24
IUPAC navn 1-cyclohexyloxycarbonyloxyethyl (6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;dihydrochloride
Molekylvægt (g/mol) 768.76
Formel vægt 767.1512