Få mere at vide
Cefotiam Hexetil Hydrochloride (mixture of Diastereomers), TRC
Artikelnummer. 30364397
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30364397 10 mg
1 options
Artikelnummer. 30364397 Leverandør Toronto Research Chemicals Leverandørnr. TRCC24300010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cefotiam Hexetil hydrochloride (mixture of diastereomers)
CAS 95789-30-3
Molekylær formel C27 H37 N9 O7 S3 . 2 Cl H
Mængde 10 mg
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[2-(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, hydrochloride (1:2), (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, dihydrochloride, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, dihydrochloride, [6R-(6α,7β)]-, Cefotiam hexetil hydrochloride, Pansporin T, SCE 2174
InChI formel InChI=1S/C27H37N9O7S3.2ClH/c1-15(42-27(40)43-18-7-5-4-6-8-18)41-24(39)21-16(13-46-26-31-32-33-35(26)10-9-34(2)3)12-44-23-20(22(38)36(21)23)30-19(37)11-17-14-45-25(28)29-17;;/h14-15,18,20,23H,4-13H2,1-3H3,(H2,28,29)(H,30,37);2*1H/t15?,20-,23-;;/m1../s1
SMIL Cl.Cl.CC(OC(=O)OC1CCCCC1)OC(=O)C2=C(CSc3nnnn3CCN(C)C)CS[C@@H]4[C@H](NC(=O)Cc5csc(N)n5)C(=O)N24
IUPAC navn 1-cyclohexyloxycarbonyloxyethyl (6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;dihydrochloride
Molekylvægt (g/mol) 768.76
Formel vægt 767.1512