missing translation for 'onlineSavingsMsg'
Learn More

Cefotaxime, 98%, ACROS Organics™

Brand:  Acros Organics 455270050

Additional Details : CAS Number : 63527-52-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15482154

  • 1140.32 DKK / 5g


Estimated Shipment
12-10-2020
Add to basket

Specifications

Chemical Identifiers

63527-52-6
455.46
53850837
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)C(=NOC)C3=CSC(=N3)N)SC1)C(=O)O
C16H17N5O7S2
GPRBEKHLDVQUJE-YGRLFVJLSA-N
(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

Specifications

Cefotaxime
Authentic
GPRBEKHLDVQUJE-YGRLFVJLSA-N
(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
53850837
5g
63527-52-6
C16H17N5O7S2
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)C(=NOC)C3=CSC(=N3)N)SC1)C(=O)O
455.46
455.47
Videos
Documents