Få mere at vide
Cefminox Sodium Heptahydrate, TRC
Artikelnummer. 30353316
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353316 5 g
30358166 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30353316 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2431005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75498-96-3
Molekylær formel C16H34N7NaO14S3
Molekylvægt (g/mol) 667.66
Synonym (6R,7S)-7-[[2-[[(2S)-2-Amino-2-carboxyethyl]thio]acetyl]amino]-7-methoxy-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Sodium Salt Heptahydrate, Cefminox Sodium Salt Heptahydrate, Meicelin Heptahydrate
IUPAC navn sodium;(6R,7S)-7-[[2-[(2S)-2-amino-2-carboxyethyl]sulfanylacetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMIL CN1C(=NN=N1)SCC2=C(N3[C@@H]([C@@](C3=O)(NC(=O)CSCC(C(=O)O)N)OC)SC2)C(=O)[O-].[Na+]
Tekniske data
Kemisk navn eller materiale Cefminox Sodium Heptahydrate
Mængde 5 g
InChI formel InChI=1S/C16H21N7O7S3.Na/c1-22-15(19-20-21-22)33-4-7-3-32-14-16(30-2,13(29)23(14)10(7)12(27)28)18-9(24)6-31-5-8(17)11(25)26;/h8,14H,3-6,17H2,1-2H3,(H,18,24)(H,25,26)(H,27,28);/q;+1/p-1/t8-,14-,16+;/m1./s1
Formel vægt 541.0484