Få mere at vide
Cefmetazole sodium, MedChemExpress
Beskrivelse
Cefmetazole sodium binds to penicillin binding proteins (PBPs), resulting in interfering bacterial cell wall biosynthesis. Cefmetazole sodium is used for the research of gynecologic, intraabdominal, urinary tract, respiratory tract and skin and soft tissue infections.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cefmetazole sodium |
| Fysisk form | Solid |
| CAS | 56796-39-5 |
| Farve | White |
| Mængde | 100 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C15H16N7NaO5S3 |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Synonym | Sodium cefmetazole |
| Opløselighedsinformation | DMSO : 250 mg/mL (506.57 mM; Need ultrasonic) ∣H2O : 100 mg/mL (202.63 mM; Need ultrasonic) |
| Vis mere |