Få mere at vide
Cefepime hydrochloride monohydrate, Thermo Scientific™
Cefepime hydrochloride monohydrate, CAS # 123171-59-5, is a hydrochloride salt of the antibiotic derived from Acremonium fungus.
Brand: Thermo Scientific Alfa Aesar J66237.06
| Mængde | 5g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Cefepime hydrochloride is a fourth-generation cephalosporin antibiotic that works against most antibiotic resistant strains of bacteria
• Cefepime hydrochloride is a penicillin binding protein inhibitor that prevents peptidoglycan cross-linkage and leads to cell wall weakening and lysis
Application
• Cefepime hydrochloride monohydrate can be used for the laboratory study of microbial physiology and antibiotic resistance
• Cefepime has broad-spectrum activity; it is appropriate for research on bacteria responsible for resistant infections and physiological studies
Kemiske identifikatorer
| 123171-59-5 | |
| 572.497 | |
| LRAJHPGSGBRUJN-OMIVUECESA-O | |
| (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride |
| C19H29Cl2N6O6S2+ | |
| MFCD01687679 | |
| 129893984 | |
| C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl |
Tekniske data
| Cefepime hydrochloride monohydrate | |
| White to Yellow | |
| Light sensitive | |
| MFCD01687679 | |
| LRAJHPGSGBRUJN-OMIVUECESA-O | |
| (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride | |
| 129893984 |
| 123171-59-5 | |
| 5g | |
| C19H29Cl2N6O6S2+ | |
| Soluble in DMSO | |
| C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl | |
| 572.497 | |
| 571.5 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.