Få mere at vide
Cefazolin Sodium Salt, TRC
Artikelnummer. 17953805
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17953805 250 mg
17940935 1 g
17972515 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17953805 Leverandør Toronto Research Chemicals Leverandørnr. TRCC242500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27164-46-1
Molekylær formel C14 H13 N8 O4 S3 . Na
Molekylvægt (g/mol) 476.49
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-, monosodium salt, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-, monosodium salt, (6R-trans)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[2-(1H-tetrazol-1-yl)acetamido]-, monosodium salt (8CI), 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-7-[2-(1H-tetrazol-1-yl)-acetamido]-3-cephem-4-carboxylic acid sodium salt, 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[2-(1H-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, Acef, Ancef, Atirin, Azolin, Biazolina, Bor-Cefazol, Cefacidal, Cefamedin, Cefazil, Cefazina, Cefazoliin, Cefazolin sodium, Cefazolin sodium salt, Cefazoline sodium, Cefazorin DC, Cephazolin sodium, Cephazolin sodium salt, Firmacef, Gramaxin, Iespor, Kefzol, Lampocef, Liviclina, Monosodium cafazolin, NSC 291561, Reflin, SKF 41558, Sodium CEZ, Sodium cefazolin, Sodium cephazolin, Totacef, Zolicef
IUPAC navn sodium;(6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMIL [Na+].Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)Cn4cnnn4)C3=O)C(=O)[O-])s1
Tekniske data
Kemisk navn eller materiale Cefazolin Sodium
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C14H14N8O4S3.Na/c1-6-17-18-14(29-6)28-4-7-3-27-12-9(11(24)22(12)10(7)13(25)26)16-8(23)2-21-5-15-19-20-21;/h5,9,12H,2-4H2,1H3,(H,16,23)(H,25,26);/q;+1/p-1/t9-,12-;/m1./s1
Formel vægt 476.012
Procent renhed >95%