Få mere at vide
Cefazolin Lactone, TRC
Artikelnummer. 30134932
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134932 50 mg
30006916 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134932 Leverandør Toronto Research Chemicals Leverandørnr. TRCC26256550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1172998-53-6
Synonym N-[(5aR,6R)-1,4,5a,6-Tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl]-1H-tetrazole-1-acetamide
Tekniske data
Format Neat
Molekylvægt (g/mol) 322.2