Få mere at vide
Cefalonium Dihydrate, Mikromol™
Artikelnummer. 17940556
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
17940556 250 mg
1 options
Artikelnummer. 17940556 Leverandør Mikromol Leverandørnr. MM3169.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cefalonium Dihydrate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cefalonium Dihydrate
CAS 1385046-35-4
Analyt- eller komponentnavne Cefalonium Dihydrate
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C20 H18 N4 O5 S2 . 2 H2 O
Certificeringer/compliance ISO 9001
Synonym 4-Carbamoyl-1-[[2-carboxy-8-oxo-7-[2-(2-thienyl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium hydroxide, inner salt, 4-(Aminocarbonyl)-1-[[(6R,7R)-2-carboxy-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium, inner salt, (6R-trans)-4-(Aminocarbonyl)-1-[[2-carboxy-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium hydroxide, inner salt, 4-Carbamoyl-1-[[2-carboxy-8-oxo-7-[2-(2-thienyl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium hydroxide, inner salt, 3-(4-Carbamoylpyridylmethyl)-8-oxo-7-[alpha-(thien-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(alpha-Thienylacetamido)-3-(4-carbamoylpyridinomethyl)-3-cephem-4-carboxylic acid, Cefalonium, Cepalonium, Cephalonium, Cepravin DC, Cepravin Dry Cow, (6R-trans)-4-(Aminocarbonyl)-1-[[2-carboxy-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinium , inner salt
InChI formel InChI=1S/C20H18N4O5S2.2H2O/c21-17(26)11-3-5-23(6-4-11)9-12-10-31-19-15(18(27)24(19)16(12)20(28)29)22-14(25)8-13-2-1-7-30-13;;/h1-7,15,19H,8-10H2,(H3-,21,22,25,26,28,29);2*1H2/t15-,19-;;/m1../s1
SMIL O.O.NC(=O)c1cc[n+](CC2=C(N3[C@H](SC2)[C@H](NC(=O)Cc4cccs4)C3=O)C(=O)[O-])cc1
IUPAC navn (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;dihydrate
Molekylvægt (g/mol) 494.54
Mængde 250 mg
Formel vægt 494.093 g/mol