missing translation for 'onlineSavingsMsg'
Learn More

Cefadroxil-d4 (Major), TRC

Product Code. 30351433
Change view
Click to view available options
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30351433 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30351433 Supplier Toronto Research Chemicals Supplier No. TRCC2357521MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Cefadroxil-d4
CAS 1426174-38-0
Renhedsgrad noter HPLC
Molekylær formel C16H13D4N3O5S • H2O
Mængde 1 mg
Synonym (6R,7R)-7-[[(2R)-Amino-(4-hydroxyphenyl-d4)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Monohydrate, p-Hydroxycephalexine-d4 Monohydrate, BL-S578-d4, MJF-11567-3-d4, Baxan-d4
InChI formel InChI=1S/C16H17N3O5S.H2O/c1-7-6-25-15-11(14(22)19(15)12(7)16(23)24)18-13(21)10(17)8-2-4-9(20)5-3-8;/h2-5,10-11,15,20H,6,17H2,1H3,(H,18,21)(H,23,24);1H2/t10-,11-,15-;/m1./s1/i2D,3D,4D,5D;
SMIL CC1=C(C(O)=O)N2C([C@@]([H])(NC([C@H](N)C3=C([2H])C([2H])=C(O)C([2H])=C3[2H])=O)[C@@]2([H])SC1)=O.O
IUPAC navn (6R,7R)-7-[[(2R)-2-amino-2-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate
Molekylvægt (g/mol) 367.411802
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.