missing translation for 'onlineSavingsMsg'
Få mere at vide

Cefaclor Monohydrate, TRC

Artikelnummer. 30132640
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132640 1 g
30137453 250 mg
17933315 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30132640 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2352501G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 70356-03-5
Molekylær formel C15 H14 Cl N3 O4 S . H2 O
Molekylvægt (g/mol) 385.82
Synonym Cefaclor Monohydrate, (6R,7R)-7-[[(2R)-Aminophenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monohydrate, 7-(D-2-amino-2-phenylacetamido)-3-chloro-3-cephem-4-carboxylic acid monohydrate
IUPAC navn (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate
SMIL O.N[C@@H](C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)Cl)c3ccccc3

Tekniske data

Kemisk navn eller materiale Cefaclor Monohydrate
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C15H14ClN3O4S.H2O/c16-8-6-24-14-10(13(21)19(14)11(8)15(22)23)18-12(20)9(17)7-4-2-1-3-5-7;/h1-5,9-10,14H,6,17H2,(H,18,20)(H,22,23);1H2/t9-,10-,14-;/m1./s1
Formel vægt 385.0499
Procent renhed >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.