missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Cefaclor Monohydrate, Mikromol™
Description
Cefaclor Monohydrate
Specifications
Specifications
| Kemisk navn eller materiale | Cefaclor Monohydrate |
| CAS | 70356-03-5 |
| Analyt- eller komponentnavne | Cefaclor Monohydrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H14 Cl N3 O4 S . H2 O |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Cefaclor Monohydrate, (6R,7R)-7-[[(2R)-Aminophenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monohydrate, 7-(D-2-amino-2-phenylacetamido)-3-chloro-3-cephem-4-carboxylic acid monohydrate |
| InChI formel | InChI=1S/C15H14ClN3O4S.H2O/c16-8-6-24-14-10(13(21)19(14)11(8)15(22)23)18-12(20)9(17)7-4-2-1-3-5-7;/h1-5,9-10,14H,6,17H2,(H,18,20)(H,22,23);1H2/t9-,10-,14-;/m1./s1 |
| SMIL | O.N[C@@H](C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)Cl)c3ccccc3 |
| IUPAC navn | (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?