Få mere at vide
Cefaclor Monohydrate, LoGiCal
Artikelnummer. 30463889
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463889 10 mg
1 options
Artikelnummer. 30463889 Leverandør LoGiCal Leverandørnr. LGCFOR0326.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cefaclor Monohydrate
CAS 70356-03-5
Analyt- eller komponentnavne Cefaclor Monohydrate
Forsendelsestilstand Room Temperature
Molekylær formel C15 H14 Cl N3 O4 S . H2 O
Certificeringer/compliance ISO 9001
Synonym Cefaclor Monohydrate, (6R,7R)-7-[[(2R)-Aminophenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monohydrate, 7-(D-2-amino-2-phenylacetamido)-3-chloro-3-cephem-4-carboxylic acid monohydrate
InChI formel InChI=1S/C15H14ClN3O4S.H2O/c16-8-6-24-14-10(13(21)19(14)11(8)15(22)23)18-12(20)9(17)7-4-2-1-3-5-7;/h1-5,9-10,14H,6,17H2,(H,18,20)(H,22,23);1H2/t9-,10-,14-;/m1./s1
Løsningstype Neat
SMIL O.N[C@@H](C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)Cl)c3ccccc3
IUPAC navn (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate
Industry Type Clinical Testing
Molekylvægt (g/mol) 385.82
Mængde 10 mg
Formel vægt 385.0499 g/mol