Få mere at vide
Tocris Bioscience™ CDPPB
GREENER_CHOICE
Artikelnummer. 18095932
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18095932 50 mg
18095622 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18095932 Leverandør Tocris Bioscience™ Leverandørnr. 3235/50
Positive allosteric modulator of mGlu5 receptors
Brain penetrant, selective positive allosteric modulator at the mGlu5 receptor (EC50 values are 10 and 20 nM for human and rat receptors respectively). Antipsychotic; reverses amphetamine-induced locomotor activity and amphetamine-induced deficits in prepulse inhibition in rats.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 781652-57-1
Molekylær formel C23H16N4O
Tekniske data
Kemisk navn eller materiale CDPPB
Mængde 50 mg
Formel vægt 364.4
Renhed >98%
Molekylvægt (g/mol) 364.408