missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CDKN2C Recombinant Protein

2665.00 DKK - 4040.00 DKK

Specifications

Accession Number AAH16173
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 1031
Molecular Weight (g/mol) 44.6
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16123071
View Documents Promotion Details
Abnova™
H00001031-P01.10UG
10 ug
2665.00 DKK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16133071
View Documents Promotion Details
Abnova™
H00001031-P01.25UG
25 ug
4040.00 DKK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH16173
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
44.6
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDKN2C
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
1031
CDKN2C (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEPWGNELASAAARGDLEQLTSLLQNNVNVNAQNGFGRTALQVMKLGNPEIARRLLLRGANPDLKDRTGFAVIHDAARAGFLDTLQTLLEFQADVNIEDNEGNLPLHLAAKEGHLRVVEFLVKHMASNVGHRNHKGDTACDLARLYGRNEVVSLMQANGAGGATNLQ
INK4C/p18/p18-INK4C
CDKN2C
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Product Certifications