Learn More
CDK2 Monoclonal Antibody (1A6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517052
Additional Details : Weight : 0.01000kg
Description
MA5-17052 targets CDK2 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Mouse samples. The MA5-17052 immunogen is purified recombinant fragment of human CDK2 expressed in E. Coli. MA5-17052 detects CDK2 which has a predicted molecular weight of approximately 33.9kDa.
CDK2/cyclinO is a member of the cyclin dependent protein kinase family complexed to CyclinO. CyclinO was used as a specific protein isoform name for the UNG gene which is localized exclusively in the nucleus. In addition, CyclinO was reported to shows significant uracil-DNA glycosylase activity. CDK2/cyclinO protein levels increase during G1 phase and the protein is turned over during the course of cell cycle. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.Specifications
CDK2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P24941, P97377 | |
CDK2 | |
Purified recombinant fragment of human CDK2 expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1A6 | |
Unconjugated | |
CDK2 | |
A630093N05Rik; CDC28; CDC2A; cdc2-related protein kinase; CDK1; CDK2; CDKN2; Cell division protein kinase 2; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 2; EC 2.7.11.22; EC 2.7.11.23; kinase Cdc2; MPF; p33 protein kinase; p33(CDK2); p33CDK2; p34 protein kinase | |
Mouse | |
Protein G | |
RUO | |
1017, 12566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |