missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD96 Antibody (6A6), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-12037JF549
Additional Details : Weight : 0.00970kg
Description
CD96 Monoclonal antibody specifically detects CD96 in Mouse samples. It is validated for Flow Cytometry, ELISASpecifications
CD96 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
10225 | |
Store at 4C in the dark. | |
IgG λ |
Flow Cytometry, ELISA | |
6A6 | |
Flow Cytometry, ELISA | |
CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile | |
This antibody was raised by immunising rats with murine CD96. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |