Learn More
CD95 (APO-1/Fas) Monoclonal Antibody (15A7), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1332.00 DKK - 3150.00 DKK
Specifications
Antigen | CD95 (APO-1/Fas) |
---|---|
Clone | 15A7 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15855503
|
Affymetrix eBioscience
62-0951-82 |
100 μg |
3150.00 DKK
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15845503
|
Affymetrix eBioscience
62-0951-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 15A7 monoclonal antibody reacts with mouse CD95 (Fas, Apo-1), a 40-50 kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, activated lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. 15A7 reacts with mouse CD95 in BALB/c strain and a bit more weakly in C57Bl/6 strain. 15A7 does not induce apoptosis of Fas-expressing cells. Applications Reported: This 15A7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 15A7 antibody has been tested by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 105 to 108 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.
CD95 (APO-1/Fas) Monoclonal antibody specifically detects CD95 (APO-1/Fas) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD95 (APO-1/Fas) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P25446 | |
FAS | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
15A7 | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Mouse | |
14102 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only