Learn More
IBA Lifesciences CD9 Fab-Strep
Low affinity Fab fragment fused to a Twin-Strep-tag™ and specific for human CD9
Brand: IBA Lifesciences 6-8019-150
Additional Details : Weight : 0.00600kg
Description
- CD9 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™
- Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
- Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to exosomes
- For traceless affinity isolation of exosomes (Fab-TACS™ for exosomes), Fab-Streps are combined with Strep-Tactin™ TACS Agarose Columns
Specifications
Lyophilized | |
Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation | |
Human |
For Research Use Only.