missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD8 Fab-Strep
Low affinity Fab fragment fused to a Twin-Strep-tag™
216.00 DKK
Specifications
Form | Lyophilized |
---|---|
For Use With (Application) | Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation |
Product Code | Brand | Species | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Species | Price | Quantity & Availability | |||||
17237600
|
IBA Lifesciences
6-8003-150 |
Human |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17227610
|
IBA Lifesciences
6-8505-150 |
Mouse |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- CD8 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™.
- Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
- Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells.
- Fab-Streps were developed for Traceless Affinity Cell Selection (Fab-TACS™), which is based on our Strep-tag™ technology.
Specifications
Lyophilized | |
Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation |
For Research Use Only.