Få mere at vide
Invitrogen™ CD79b Monoclonal Antibody (HM79-11)

Armenian Hamster Monoclonal Antibody
Brand: Invitrogen™ MA180066
Beskrivelse
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Hamster anti Mouse CD79b antibody, clone HM79-11 recognizes murine CD79 beta, expressed by B cells as part of the B cell receptor complex (immunoglobulin and the CD79 alpha/beta heterodimer).
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
Tekniske data
| CD79b | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P15530 | |
| Cd79b | |
| CD79 alpha/CD79 beta heterodimer purified from WEH1231 B cells. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| HM79-11 | |
| Unconjugated | |
| Cd79b | |
| AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7 | |
| Armenian Hamster | |
| Protein G | |
| RUO | |
| 15985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.