missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD68/SR-D1 Antibody (SPM130), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34736AF532
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human, Feline, Monkey, Rabbit, Canine (Negative), Chicken (Negative), Porcine (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Tekniske data
| CD68/SR-D1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD68 | |
| This CD68/SR-D1 Antibody (SPM130) was developed against subcellular fraction of human alveolar macrophages | |
| 0.1 mL | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| This antibody recognizes a glycoprotein of 110kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia and also reacts with rare cases of true histiocytic neoplasia. Lymphomas are negative or show few granules. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM130 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Feline, Primate, Canine (Negative), Chicken (Negative), Porcine (Negative) | |
| IgG1 κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion