Få mere at vide
CD58/LFA-3 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF1689-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD58/LFA-3 Polyclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only