missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD53 Antibody (161-2 (53/2)), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP3-12076AF532
Additional Details : Weight : 0.00970kg
Description
CD53 Monoclonal antibody specifically detects CD53 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
CD53 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG2a κ |
Flow Cytometry, Immunofluorescence | |
161-2 (53/2) | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
This antibody was raised by immunizng mice with stimulated human leukocytes. | |
0.1 mL | |
Immunology | |
963 | |
Store at 4°C in the dark. |