missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Rabbit Monoclonal Antibody, Epredia™
Obtain accurate and reproducible results in immunohistochemistry and western blotting procedures with Epredia™ CD5, Rabbit Monoclonal Antibody.
1325.00 DKK - 5880.00 DKK
Specifications
Antigen | CD5 |
---|---|
Clone | SP19 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12693757
|
Epredia
RM-9119-S0 |
Concentrated | 100 μL |
1325.00 DKK
100µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12603767
|
Epredia
RM-9119-S1 |
Concentrated | 500 μL |
4575.00 DKK
500µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12683757
|
Epredia
RM-9119-S |
Concentrated | 1 mL |
5880.00 DKK
1mL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12663757
|
Epredia
RM-9119-R7 |
Prediluted | 7 mL |
3265.00 DKK
7mL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12673757
|
Epredia
RM-9119-RQ |
Ready-to-Use | 12 mL |
4810.00 DKK
12mL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
CD5, a transmembrane protein, is found on 95%of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.Host Species: Rabbit
Clone: SP19
Species Reactivity: Human. Others not tested.
Epitope: Intracellular region
Immunogen: Synthetic peptide from the intracellular region of CD5
Positive Control: Raji cells, Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
CD5 | |
Western Blot | |
Unconjugated | |
IVD | |
Synthetic peptide from the intracellular region of CD5 |
SP19 | |
Monoclonal | |
Rabbit | |
Hematopoietic | |
Primary |