Få mere at vide
CD45RA Mouse anti-Rat, BUV661, Clone: OX-33, BD Biosciences™
Beskrivelse
CD45RA Monoclonal antibody specifically detects CD45RA in Rat samples. It is validated for Flow Cytometry
Tekniske data
Tekniske data
| Antigen | CD45RA |
| Applikationer | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | OX-33 |
| Konjugeret | Brilliant Ultraviolet 661 |
| Formulering | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Ptprc; Lca; Leucocyte common antigen |
| Værtsarter | Mouse |
| Immunogen | Leukocyte common antigen purified from rat splenocytes |
| Mængde | 50 μg |
| Vis mere |