Learn More
CD44 Monoclonal Antibody (IM7), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1188.00 DKK - 3040.00 DKK
Specifications
Antigen | CD44 |
---|---|
Clone | IM7 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15875493
|
Affymetrix eBioscience
62-0441-82 |
100 μg |
3040.00 DKK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15865493
|
Invitrogen
62-0441-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The IM7 monoclonal antibody reacts with all isoforms of mouse CD44 (Pgp-1). CD44 is expressed by hematopoietic and non-hematopoietic cells. Bone marrow myeloid cells and memory T cells highly express this antigen and peripheral B and T cells can upregulate the expression of CD44. CD44 functions as an adhesion molecule through its binding to hyaluronate, an extracellular matrix component. Applications Reported: This IM7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This IM7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions.
CD44 Monoclonal antibody specifically detects CD44 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
CD44 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
12505, 960 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
IM7 | |
Flow Cytometry | |
Super Bright 436 | |
Rat | |
Human, Mouse | |
CD44 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD44 |
For Research Use Only