missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43 Mouse anti-Human, FITC, Clone: L10, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD4301
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD43 (leukosialin, sialophorin) is a transmembrane mucin-like protein with high negative charge, expressed on the surface of most hematopoietic cells. CD43 contributes to a repulsive barrier that interferes with cellular adhesion, however, in certain cases also promotes leukocyte aggregation. By interaction with actin-binding proteins ezrin and moesin CD43 plays a regulatory role in remodeling T-cell morphology and regulates cell-cell interactions during lymphocyte traffic. CD43 signaling both enhances LFA-1 adhesiveness and counteracts LFA-1 induction via other receptors. Expression of CD43 causes induction of functionally active tumor suppressor p53 protein, but in case of p53 and ARF deficiency CD43 promotes tumor proliferation and viability. CD43 appears to be an important modulator of leukocyte functions. Diseases associated with CD43 dysfunction include Wiscott-Aldrich Syndrome and Adenoid Basal Cell Carcinoma.Specifications
CD43 | |
Monoclonal | |
FITC | |
SPN | |
CD43, GALGP, GPL115, LSN, galactoglycoprotein, leukocyte sialoglycoprotein, leukosialin, sialophorin gpL115, leukosialin, CD43 | |
Mouse | |
Purified | |
ASR | |
6693 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
L10 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P16150 | |
SPN | |
Human CD43 | |
0.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |