Få mere at vide
CD42b/GPIb alpha Antibody (HIP1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-511AF488
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human, Mouse, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Tekniske data
| CD42b/GPIb alpha | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody HIP1 reacts with CD42b (GPIb alpha), a 135-145 kDa membrane glycoprotein expressed on platelets and megakaryocytes. CD42b and CD42c (GPIb beta) are composed in a disulfide linked heterodimer (CD42b/c; 160 kDa); CD42b/c forms a noncovalent complex with CD42a and CD42d. HLDA IV; WS Code P 40 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| HIP1 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| GP1BA | |
| Peripheral blood mononuclear cells of a patient suffering with CLL. | |
| 0.1 mL | |
| Immunology | |
| 2811 | |
| Human, Mouse, Primate | |
| IgG1 |
Korrektion af produktindhold
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion