missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD42b/GPIb alpha Antibody (486805) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4067JF635
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD42b/GPIb alpha Monoclonal antibody specifically detects CD42b/GPIb alpha in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Tekniske data
| CD42b/GPIb alpha | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
| Mouse myeloma cell line NS0-derived recombinant human CD42b/GPIb alpha , His17-Leu505, Accession # P07359 | |
| 0.1 mL | |
| Immunology | |
| 2811 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Western Blot, CyTOF | |
| 486805 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion