Beskrivelse
CD41/CD61 Monoclonal antibody specifically detects CD41/CD61 in Human samples. It is validated for Immunohistochemistry
Tekniske data
Tekniske data
| Antigen | CD41/CD61 |
| Applikationer | Immunohistochemistry |
| Klassifikation | Monoclonal |
| Klon | BAG 75E5 |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Fortynding | Immunohistochemistry |
| Formulering | PBS |
| Gene Alias | CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK |
| Værtsarter | Mouse |
| Vis mere |