missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Abnova™ CD3D 293T Cell Transient Overexpression Lysate (Denatured)
Tekniske data
Tekniske data
| Cellelinje | 293T |
| Til brug med (applikation) | Western Blot |
| Oversigt | CD3D 293T Cell Transient Overexpression Lysate(Denatured) |
| Genadgangsnr. | NM_000732 |
| Molekylvægt (g/mol) | Theoretical M.W. 18.9kDa |
| Plasmid | pCMV-CD3D full-length |
| Kvalitetskontrol test | Transient overexpression cell lysate was tested with anti-CD3D Purified antibody (Mfr. No. H00000915-B01P) by Western Blot. |
| Mængde | 100 μL |
| Opbevaringsbuffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Værtsarter | Human |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- 100UL CD3D 293T Cell Transient Overexpression Lysa
- Advarsel
- Hudsensibilisering Kategori 1
- H317-Kan forårsage allergisk hudreaktion.
- P261-Undgå indånding af pulver/røg/gas/tåge/damp/spray.
- P272-Tilsmudset arbejdstøj bør ikke fjernes fra arbejdspladsen.
- P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
- MIXTURE LIST-Contain: Mercaptoethanol
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?