missing translation for 'onlineSavingsMsg'
Learn More

CD39 Rat anti-Mouse, Alexa Fluor® 700, Clone: 24DMS1, eBioscience™

Rat Monoclonal Antibody

Brand:  Invitrogen 56-0391-80

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15855463

  • 1100.00 DKK / 25µg


Estimated Shipment date
from Supplier 29-05-2019
Add to basket

Description

Description

CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.
Specifications

Specifications

CD39
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide
P55772
Ectonucleoside Triphosphate Diphosphohydrolase 1, Entpd1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
24DMS1
Alexa Fluor® 700
Entpd1
Liquid
Entpd1
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12495
Videos