missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CD38 (Human) Recombinant Protein

Human CD38 full-length ORF ( AAH07964, 1 a.a. - 300 a.a.) recombinant protein with GST-tag at N-terminal.

2735.00 DKK - 4145.00 DKK

Specifications

Accession Number AAH07964
Gene ID (Entrez) 952
Name CD38 molecule
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16122861
View Documents
Abnova™
H00000952-P01.10ug
10 ÎĽg
2735.00 DKK
10µg
Please to purchase this item. Need a web account? Register with us today!
16132861
View Documents
Abnova™
H00000952-P01.25ug
25 ÎĽg
4145.00 DKK
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Sequence: MANCEFSPVSGDKPCCRLSRRAQLCLGVSILVLILVVVLAVVVPRWRQQWSGPGTTKRFPETVLARCVKYTEIHPEMRHVDCQSVWDAFKGAFISKHPCNITEEDYQPLMKLGTQTVPCNKILLWSRIKDLAHQFTQVQRDMFTLEDTLLGYLADDLTWCGEFNTSKINYQSCPDWRKDCSNNPVSVFWKTVSRRFAEAACDVVHAMLNGSRSKIFDKNSTFGSVEVHNLQPEKVQTLEAWVIHGGREDSRDLCQDPTIKELESIISKRNIQFSCKNIYRPDKFLQCVKNPEDSSCTSEI
Specifications

Specifications

AAH07964
CD38 molecule
125% SDS-PAGE Stained with Coomassie Blue
T10
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
952
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD38
GST
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ CD38 (Human) Recombinant Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.