Få mere at vide
Abnova™ CD38 (Human) Recombinant Protein
Artikelnummer. 16132861
Mængde:
10 μg
25 μg
Pakningsstørrelse:
10µg
25µg
Artikelnummer. 16132861
Brand: Abnova™ H00000952P01.25ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Human CD38 full-length ORF ( AAH07964, 1 a.a. - 300 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MANCEFSPVSGDKPCCRLSRRAQLCLGVSILVLILVVVLAVVVPRWRQQWSGPGTTKRFPETVLARCVKYTEIHPEMRHVDCQSVWDAFKGAFISKHPCNITEEDYQPLMKLGTQTVPCNKILLWSRIKDLAHQFTQVQRDMFTLEDTLLGYLADDLTWCGEFNTSKINYQSCPDWRKDCSNNPVSVFWKTVSRRFAEAACDVVHAMLNGSRSKIFDKNSTFGSVEVHNLQPEKVQTLEAWVIHGGREDSRDLCQDPTIKELESIISKRNIQFSCKNIYRPDKFLQCVKNPEDSSCTSEI
Tekniske data
Adgangsnummer AAH07964
Gen-id (Entrez) 952
Navn CD38 molecule
Fremstillingsmetode Wheat germ expression system
Kvalitetskontrol test 125% SDS-PAGE Stained with Coomassie Blue
Mængde 25 μg
Opbevaringskrav Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias T10
Gen symbol CD38
Arter Wheat Germ (in vitro)
Protein tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer