missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD34 Antibody (My10) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
2155.00 DKK - 4485.00 DKK
Tekniske data
| Antigen | CD34 |
|---|---|
| Klon | My10 |
| Fortynding | Western Blot, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applikationer | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18243043
|
Novus Biologicals
NBP2-62554-0.025MG |
0.025 mg |
2155.00 DKK
25µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18668867
|
Novus Biologicals
NBP2-62554 |
0.2 mg |
4485.00 DKK
0.20mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation.Tekniske data
| CD34 | |
| Western Blot, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| PBS | |
| 947 | |
| KG-1a Human myeloblastic leukemic cell line (immature human myeloid cell line) from a patient with nonlymphocytic leukemia | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| My10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| CD34 | |
| IgG κ | |
| Human CD34, an ∼105 - 120 kDa type 1 transmembrane glycoprotein expressed on the surface of most human hematopoietic stem and progenitor cells (HSPCs) as well as on mesenchymal stem cells, embryonic fibroblasts, endothelial cells, neurons, and some tumor cell lines. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel