Få mere at vide
CD326 (EpCAM) Mouse, anti-Human, Unconjugated, Clone: 323/A3, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60146
Encompass_Preferred
Produktkode 17169181
  • 3856.95 DKK / 100µg
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
  • The 323/A3 antibody reacts with the human epithelial cell adhesion molecule (EpCAM or CD326), an ∼40 kDa type I transmembrane glycoprotein which functions as a homophilic and Ca++-independent adhesion molecule
  • EpCAM has roles in several cellular processes, including signaling, migration, proliferation, and differentiation
  • It is expressed early during erythrogenesis and has been identified as a stem cell marker
  • The EpCAM protein is found on the basolateral membrane of most normal epithelial cells and is highly expressed in bone marrow, colon, and lung
  • EpCAM is not expressed on mesothelial cells or mesotheliomas, and so is widely used as a diagnostic marker to distinguish mesothelioma and carcinoma cells, as well as serving as a target for immunotherapeutic treatment of carcinomas
Tekniske data
Tekniske data
CD326 (EpCAM)
Monoclonal
Unconjugated
100 μg
Human
Mammary Cells
Immunohistochemistry, Flow Cytometry, Immunocytochemistry
323/A3
Mouse
Primary
IgG1 κ
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.