Learn More
CD303a Monoclonal Antibody (201A), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1052.00 DKK - 3065.00 DKK
Specifications
Antigen | CD303a |
---|---|
Clone | 201A |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15538676
|
Affymetrix eBioscience
25-9818-42 |
100 Tests |
3065.00 DKK
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15528676
|
Affymetrix eBioscience
25-9818-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This 201A monoclonal antibody reacts with human CD303a, which is also known as CLEC4C or Blood Dendritic Cell Antigen 2 (BDCA-2). This type II transmembrane protein is expressed almost exclusively on plasmacytoid dendritic cells, which secrete type I interferon during antiviral and antitumor responses. Studies have demonstrated that CD303a binds asialo-galactosyl-oligosaccharides on invading pathogens, resulting in the activation of signaling pathways that lead to CD303a internalization and downregulation of interferon production. Applications Reported: This 201A antibody has been reported for use in flow cytometric analysis. Applications Tested: This 201A antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD303a Monoclonal antibody specifically detects CD303a in Human samples. It is validated for Flow Cytometry.Specifications
CD303a | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q8WTT0 | |
CLEC4C | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CLEC4C |
201A | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
170482 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.