missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LG/Nepmucin Antibody (102), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90215G
Additional Details : Weight : 0.00970kg
Description
CD300LG/Nepmucin Monoclonal specifically detects CD300LG/Nepmucin in Human samples. It is validated for ELISA.Specifications
CD300LG/Nepmucin | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
146894 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
102 | |
ELISA | |
CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD300LG/Nepmucin (AAH25395.1; Met1-Arg247). | |
0.1 mL | |
Primary | |
Human | |
Purified |