Få mere at vide
CD3 Mouse anti-Human, CoraLite 647, Clone: Hit3a, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-65112-100TESTS
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The CD3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD3. This antibody recognizes human antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Tekniske data
| CD3 | |
| Monoclonal | |
| 5 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| Hit3a | |
| CoraLite 647 | |
| 100 Tests | |
| 916 | |
| IgG2a |